missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl Phosphorochloridate, TRC

Product Code. 30383132
Change view
Click to view available options
Quantity:
1 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30383132 100 g
30372637 25 g
30375250 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30383132 Supplier Toronto Research Chemicals Supplier No. TRCD459400100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 2574-25-6
Molecular Formula C6 H14 Cl O3 P
Molecular Weight (g/mol) 200.6
Synonym Phosphorochloridic acid, bis(1-methylethyl) ester, Isopropyl phosphorochloridate (6CI,7CI), Phosphorochloridic acid, diisopropyl ester (8CI), (Chlorodiisopropoxy)phosphine oxide, Diisopropyl chlorophosphate, Diisopropyl phosphorochloridate, Diisopropylphosphoryl chloride, Isopropyl phosphorochloridate ((C3H7O)2ClPO)
IUPAC Name 2-[chloro(propan-2-yloxy)phosphoryl]oxypropane
SMILES CC(C)OP(=O)(Cl)OC(C)C

Specifikationer

Quantity 100 g
InChI Formula InChI=1S/C6H14ClO3P/c1-5(2)9-11(7,8)10-6(3)4/h5-6H,1-4H3
Formula Weight 200.0369
Chemical Name or Material Diisopropyl Phosphorochloridate
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.