missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl ketone, 98%

Product Code. p-7020801
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11415313

Brand: Thermo Scientific Alfa Aesar A12875.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 565-80-0 | C7H14O | 114.188 g/mol

2,4-Dimethyl-3-pentanone is used in an experimental method to measure the equilibrium isotopic fractionation factor (αeq ) for C-bound H positions adjacent to carbonyl group in ketones.It is a carbonyl compound used in an experimental method to measure the equilibrium isotopic fractionation factor (αeq ) for C-bound H positions adjacent to carbonyl group in ketones. Isobutyrone is a very useful synthetic intermediate. It is an intermediate used to prepare α-aryl ketones. It is also used in Grignard reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dimethyl-3-pentanone is used in an experimental method to measure the equilibrium isotopic fractionation factor (αeq ) for C-bound H positions adjacent to carbonyl group in ketones.It is a carbonyl compound used in an experimental method to measure the equilibrium isotopic fractionation factor (αeq ) for C-bound H positions adjacent to carbonyl group in ketones. Isobutyrone is a very useful synthetic intermediate. It is an intermediate used to prepare α-aryl ketones. It is also used in Grignard reactions.

Solubility
Not miscible or difficult to mix in water. Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from ignition sources. Keep away from electrostatic charges.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 565-80-0
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00008918
InChI Key HXVNBWAKAOHACI-UHFFFAOYSA-N
Synonym 2,4-dimethyl-3-pentanone, diisopropyl ketone, isobutyrone, isopropyl ketone, 3-pentanone, 2,4-dimethyl, diisopropylketone, unii-7aap3a50ig, iso-c3h7 2co, 7aap3a50ig, 2,4-dimethyl-pentan-3-one
PubChem CID 11271
IUPAC Name 2,4-dimethylpentan-3-one
SMILES CC(C)C(=O)C(C)C

Specifications

Melting Point -69°C
Density 0.803
Boiling Point 124°C to 125°C
Flash Point 15°C (59°F)
Refractive Index 1.399
Quantity 100 g
UN Number UN1224
Beilstein 773782
Solubility Information Not miscible or difficult to mix in water. Soluble in alcohol.
Formula Weight 114.19
Percent Purity 98%
Chemical Name or Material Diisopropyl ketone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diisopropyl ketone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contains : Diisopropyl ketone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.