missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl ketone, 98%

Product Code. 11425313
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11425313 500 g 500g
11415313 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11425313 Supplier Thermo Scientific Alfa Aesar Supplier No. A12875.36
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 565-80-0 | C7H14O | 114.188 g/mol

2,4-Dimethyl-3-pentanone is used in an experimental method to measure the equilibrium isotopic fractionation factor (αeq ) for C-bound H positions adjacent to carbonyl group in ketones.It is a carbonyl compound used in an experimental method to measure the equilibrium isotopic fractionation factor (αeq ) for C-bound H positions adjacent to carbonyl group in ketones. Isobutyrone is a very useful synthetic intermediate. It is an intermediate used to prepare α-aryl ketones. It is also used in Grignard reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dimethyl-3-pentanone is used in an experimental method to measure the equilibrium isotopic fractionation factor (αeq ) for C-bound H positions adjacent to carbonyl group in ketones.It is a carbonyl compound used in an experimental method to measure the equilibrium isotopic fractionation factor (αeq ) for C-bound H positions adjacent to carbonyl group in ketones. Isobutyrone is a very useful synthetic intermediate. It is an intermediate used to prepare α-aryl ketones. It is also used in Grignard reactions.

Solubility
Not miscible or difficult to mix in water. Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from ignition sources. Keep away from electrostatic charges.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 565-80-0
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00008918
InChI Key HXVNBWAKAOHACI-UHFFFAOYSA-N
Synonym 2,4-dimethyl-3-pentanone, diisopropyl ketone, isobutyrone, isopropyl ketone, 3-pentanone, 2,4-dimethyl, diisopropylketone, unii-7aap3a50ig, iso-c3h7 2co, 7aap3a50ig, 2,4-dimethyl-pentan-3-one
PubChem CID 11271
IUPAC Name 2,4-dimethylpentan-3-one
SMILES CC(C)C(=O)C(C)C

Specifications

Melting Point -69°C
Density 0.803
Boiling Point 124°C to 125°C
Flash Point 15°C (59°F)
Refractive Index 1.399
Quantity 500 g
UN Number UN1224
Beilstein 773782
Solubility Information Not miscible or difficult to mix in water. Soluble in alcohol.
Formula Weight 114.19
Percent Purity 98%
Chemical Name or Material Diisopropyl ketone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.