missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS | 55-91-4 |
| Melting Point | -82.0°C |
| Density | 1.0550g/mL |
| Boiling Point | 62.0°C (9.0 mmHg) |
| Assay Percent Range | 98.5% min. (GC) |
| Packaging | Ampoule |
| Molecular Formula | C6H14FO3P |
| Linear Formula | [(CH3)2CHO]2P(O)F |
| Quantity | 1 g |
| Merck Index | 15,5222 |
| Show More |
Safety and Handling
Product Identifier
- Diisopropyl fluorophosphate
- Danger
- Acute toxicity Category 1
- Acute toxicity Category 2
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H300-Fatal if swallowed.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P361-Take off immediately all contaminated clothing.
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?