missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl Ether, ReagentPlus™, 99%, Honeywell™

Product Code. p-7120390 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 L
2.5 L
4 L
Packaging:
Aluminum Bottle
Glass Bottle
Unit Size:
1 L
2.5 L
4 L
This item is not returnable. View return policy

Product Code. 15631670

Brand: Honeywell Research Chemicals 1853021L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, contains either BHT or hydroquinone as stabilizer, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-20-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD00008880
InChI Key ZAFNJMIOTHYJRJ-UHFFFAOYSA-N
Synonym diisopropyl ether, isopropyl ether, propane, 2,2'-oxybis, 2-isopropoxypropane, diisopropyl oxide, diisopropylether, ether, isopropyl, 2,2'-oxydipropane, isopropylether, bis isopropyl ether
PubChem CID 7914
IUPAC Name 2-propan-2-yloxypropane
SMILES CC(C)OC(C)C

Specifications

Melting Point -86°C
Color Colorless
Density 0.725g/mL at 25°C
Boiling Point 70°C
Explosion Limit 1 to 21% (100°F)
Flash Point −29°C (−20.2°F)
Ignition Point 827°F
Packaging Glass Bottle
Linear Formula (CH3)2CHOCH(CH3)2
Refractive Index n20/D 1.367(lit.)
Quantity 1 L
UN Number UN1159
Beilstein 1731256
Vapor Density 3.5 (vs air)
Formula Weight 102.17g/mol
Vapor Pressure 120mmHg (20°C)
Percent Purity 99%
Grade ReagentPlus™
Physical Form Liquid
Chemical Name or Material Diisopropyl Ether, Contains either BHT or hydroquinone as stabilizer
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diisopropyl ether
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H336-May cause drowsiness or dizziness.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.