missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl ether, ACS Reagent, contains either BHT or hydroquinone as stabilizer, ≥99.0%, Honeywell Riedel-de Haën™

Product Code. 15625380
Change view
Click to view available options
Quantity:
500 mL
6 x 1 L
Packaging:
Fiberboard box
Glass Bottle
Unit Size:
500mL
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15625380 500 mL Glass Bottle 500mL
15635380 6 x 1 L Fiberboard box Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15625380 Leverantör Honeywell-Riedel-de Haen Leverantörsnummer 398276500ML
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

ACS Reagent, contains either BHT or hydroquinone as stabilizer,≥99.0%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 108-20-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD00008880
InChI Key ZAFNJMIOTHYJRJ-UHFFFAOYSA-N
Synonym diisopropyl ether, isopropyl ether, propane, 2,2'-oxybis, 2-isopropoxypropane, diisopropyl oxide, diisopropylether, ether, isopropyl, 2,2'-oxydipropane, isopropylether, bis isopropyl ether
PubChem CID 7914
IUPAC Name 2-propan-2-yloxypropane
SMILES CC(C)OC(C)C

Specifikationer

Melting Point -86°C
Density 0.725g/mL at 25°C
Boiling Point 67°C to 70°C (1013 hPa)
Flash Point −29°C (−20.2°F)
Odor Odorless
Packaging Glass Bottle
Linear Formula (CH3)2CHOCH(CH3)2
Quantity 500 mL
UN Number UN1159
Beilstein 1731256
Vapor Density 3.5 (vs air)
Formula Weight 102.17g/mol
Percent Purity ≥99%
Chemical Name or Material Diisopropyl ether
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.