missing translation for 'onlineSavingsMsg'
Learn More

(-)-Diisopropyl D-tartrate, 99%

Product Code. 10626632 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 10626632

Brand: Thermo Scientific Acros 211180100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 62961-64-2 | C10H18O6 | 234.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62961-64-2
Molecular Formula C10H18O6
Molecular Weight (g/mol) 234.25
MDL Number MFCD00008876
InChI Key XEBCWEDRGPSHQH-YUMQZZPRSA-N
Synonym diisopropyl d-tartrate, 2s,3s-diisopropyl 2,3-dihydroxysuccinate, --diisopropyl d-tartrate, -diisopropyl-d-tartrate, d---tartaric acid diisopropyl ester, diisopropyl d---tartrate, butanedioic acid, 2,3-dihydroxy-, bis 1-methylethyl ester, 2s,3s, d---diisopropyl tartrate, d-dipt, --d-tartaric acid diisopropyl ester, diisoprropyl-d-tartrate
PubChem CID 112972
IUPAC Name dipropan-2-yl (2S,3S)-2,3-dihydroxybutanedioate
SMILES CC(C)OC(=O)C(C(C(=O)OC(C)C)O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 1.1100g/mL
Boiling Point 275°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula [-CH(OH)CO2CH(CH3)2]2
Refractive Index 1.4380 to 1.44
Quantity 10 g
Fieser 13,276
Specific Gravity 1.11
Specific Rotation Condition −17.25° (20°C neat)
Specific Rotation -17.25
Formula Weight 234.25
Percent Purity 99%
Physical Form Viscous Liquid
Chemical Name or Material (-)-Diisopropyl D-tartrate, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.