missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl Cyanomethylphosphonate, TRC

Product Code. 30384084
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30384084 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30384084 Supplier Toronto Research Chemicals Supplier No. TRCD5254205G
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diisopropyl Cyanomethylphosphonate
CAS 21658-95-7
Molecular Formula C8H16NO3P
Quantity 5 g
Synonym Diisopropyl Phosphonoacetonitrile, (Cyanomethyl)phosphonic Acid Diisopropyl Ester, (Cyanomethyl)phosphonic Acid Bis(1-methylethyl) Ester
InChI Formula InChI=1S/C8H16NO3P/c1-7(2)11-13(10,6-5-9)12-8(3)4/h7-8H,6H2,1-4H3
SMILES CC(C)OP(=O)(CC#N)OC(C)C
IUPAC Name 2-di(propan-2-yloxy)phosphorylacetonitrile
Molecular Weight (g/mol) 205.19
Formula Weight 205.0868
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.