missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl Cyanomethylphosphonate, TRC

Product Code. 30376634
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30376634 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30376634 Supplier Toronto Research Chemicals Supplier No. TRCD525420250MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diisopropyl Cyanomethylphosphonate
CAS 21658-95-7
Molecular Formula C8H16NO3P
Quantity 250 mg
Synonym Diisopropyl Phosphonoacetonitrile, (Cyanomethyl)phosphonic Acid Diisopropyl Ester, (Cyanomethyl)phosphonic Acid Bis(1-methylethyl) Ester
InChI Formula InChI=1S/C8H16NO3P/c1-7(2)11-13(10,6-5-9)12-8(3)4/h7-8H,6H2,1-4H3
SMILES CC(C)OP(=O)(CC#N)OC(C)C
IUPAC Name 2-di(propan-2-yloxy)phosphorylacetonitrile
Molecular Weight (g/mol) 205.19
Formula Weight 205.0868
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.