missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl cyanomethylphosphonate, 97%

Product Code. 11499687
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499687 5 g 5g
11409697 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499687 Supplier Thermo Scientific Alfa Aesar Supplier No. B22163.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 21658-95-7 | C8H16NO3P | 205.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21658-95-7
Molecular Formula C8H16NO3P
Molecular Weight (g/mol) 205.194
MDL Number MFCD00009812
InChI Key MQDIIJHQSUNBKA-UHFFFAOYSA-N
Synonym diisopropyl cyanomethyl phosphonate, diisopropyl cyanomethylphosphonate, diisopropyl cyanomethylphosphonate,, acmc-20al7r, diisopropoxyphosphinylacetonitrile, 2-di propan-2-yloxy phosphorylacetonitrile, diisopropyl cyanomethylphosphonate 5g, phosphonic acid,p-cyanomethyl-, bis 1-methylethyl ester
PubChem CID 88997
IUPAC Name 2-di(propan-2-yloxy)phosphorylacetonitrile
SMILES CC(C)OP(=O)(CC#N)OC(C)C

Specifications

Density 1.039
Boiling Point 108°C to 110°C (0.08 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4295
Quantity 5 g
UN Number UN2927
Beilstein 1946176
Formula Weight 205.19
Percent Purity 97%
Chemical Name or Material Diisopropyl cyanomethylphosphonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.