missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl Carbonate (Stabilized with Calcium Carbonate), TRC

Product Code. 30387422
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387422 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30387422 Supplier Toronto Research Chemicals Supplier No. TRCD45550025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diisopropyl Carbonate
CAS 6482-34-4
Molecular Formula C7H14O3
Quantity 25 g
Synonym Carbonic Acid Diisopropyl Ester, Bis-2-propyl Carbonate, Diisopropyl Carbonate
InChI Formula InChI=1S/C7H14O3/c1-5(2)9-7(8)10-6(3)4/h5-6H,1-4H3
SMILES CC(C)OC(=O)OC(C)C
IUPAC Name dipropan-2-yl carbonate
Molecular Weight (g/mol) 146.18
Formula Weight 146.0943
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.