missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl bromomethylphosphonate, 97%

Product Code. 11363227
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11363227 5 g 5g
11373227 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11363227 Supplier Thermo Scientific Alfa Aesar Supplier No. L13763.06
  • ONLINE EXCLUSIVE PRICE
    £46.60
    £41.65 / 5g
    Save £4.95
    10% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 98432-80-5 | C7H16BrO3P | 259.08 g/mol

Diisopropyl bromomethylphosphonate, is used as an organophosphorus intermediate. It is also used as a biochemical reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diisopropyl bromomethylphosphonate, is used as an organophosphorus intermediate. It is also used as a biochemical reagent.

Solubility
Insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98432-80-5
Molecular Formula C7H16BrO3P
Molecular Weight (g/mol) 259.08
MDL Number MFCD00236036
InChI Key JNONGIUWXHOEEN-UHFFFAOYSA-N
Synonym diisopropyl bromomethylphosphonate, bromomethylphosphonic acid diisopropyl ester, diisopropyl bromomethyl phosphonate, bromomethylphosphonate diisopropyl ester, 2-bromomethyl propan-2-yloxy phosphoryl oxypropane
PubChem CID 4366885
IUPAC Name 2-[bromomethyl(propan-2-yloxy)phosphoryl]oxypropane
SMILES CC(C)OP(=O)(CBr)OC(C)C

Specifications

Boiling Point 80°C (1 mmHg)
Refractive Index 1.4495
Quantity 5 g
UN Number UN3265
Beilstein 4740388
Solubility Information Insoluble in water.
Formula Weight 259.09
Percent Purity 97%
Chemical Name or Material Diisopropyl bromomethylphosphonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.