missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl azodicarboxylate, 94%

Product Code. p-113479 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
This item is not returnable. View return policy

Product Code. 10570812

Brand: Thermo Scientific Acros 327560050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2446-83-5 | C8H14N2O4 | 202.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X Bond Formation (Halogen), Radical initiators
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2446-83-5
Molecular Formula C8H14N2O4
Molecular Weight (g/mol) 202.21
MDL Number MFCD00008875
InChI Key VVWRJUBEIPHGQF-KTKRTIGZSA-N
Synonym z-n-propan-2-yloxy carbonyl imino propan-2-yloxy formamide
PubChem CID 56776520
IUPAC Name propan-2-yl (NZ)-N-propan-2-yloxycarbonyliminocarbamate
SMILES CC(C)OC(=O)N=NC(=O)OC(C)C

Specifications

Melting Point 3.0°C to 5.0°C
Color Orange-Yellow to Red
Density 1.0200g/mL
Boiling Point 75.0°C (0.2 mmHg)
Flash Point 106°C
Infrared Spectrum Authentic
Assay Percent Range 92% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CHO2CN=NCO2CH(CH3)2
Refractive Index 1.4180 to 1.4220
Quantity 5 g
Fieser 17,39
Merck Index 15,3217
Specific Gravity 1.02
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 202.21
Percent Purity 94%
Physical Form Liquid
Chemical Name or Material Diisopropyl azodicarboxylate, 94%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diisopropyl azodicarboxylate
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.