missing translation for 'onlineSavingsMsg'
Learn More

(+)-Diisopinocampheylchloroborane, 1.8M solution in hexanes, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10350134 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10350134 100 mL AcroSeal™ Glass Bottle 100mL
10553044 800 mL AcroSeal™ Glass Bottle 800mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10350134 Supplier Thermo Scientific Acros Supplier No. 388421000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112246-73-8 , 80-56-8 , 71-43-2 , 64742-89-8
Molecular Formula C20H34BCl
Molecular Weight (g/mol) 320.75
MDL Number MFCD00074808
InChI Key PSEHHVRCDVOTID-NAVXHOJHSA-N
Synonym +-b-chlorodiisopinocamphenylborane
PubChem CID 133124869
IUPAC Name chloro-bis[(3R,4S,5R)-4,6,6-trimethyl-3-bicyclo[3.1.1]heptanyl]borane
SMILES C[C@H]1[C@@H]2C[C@H](C[C@@H]1B(Cl)[C@H]1C[C@H]3C[C@@H]([C@@H]1C)C3(C)C)C2(C)C

Specifications

Packaging AcroSeal™ Glass Bottle
Quantity 100 mL
Solubility Information Solubility in water: reacts
Formula Weight 320.77
Chemical Name or Material (+)-Diisopinocampheylchloroborane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.