missing translation for 'onlineSavingsMsg'
Learn More

Diisopentylamine, 97%

Product Code. p-7058265
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11367655

Brand: Thermo Scientific Alfa Aesar L02297.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 544-00-3 | C10H23N | 157.301 g/mol

Intermediate for organic synthesis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Intermediate for organic synthesis

Solubility
Soluble in ethanol, ethyl ether and chloroform. Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544-00-3
Molecular Formula C10H23N
Molecular Weight (g/mol) 157.301
MDL Number MFCD00015053
InChI Key SPVVMXMTSODFPU-UHFFFAOYSA-N
Synonym diisopentylamine, diisoamylamine, 1-butanamine, 3-methyl-n-3-methylbutyl, di 3-methylbutyl amine, bis 3-methylbutyl amine, di-iso-pentylamine = di-iso-amylamine, 3-methyl-n-3-methylbutyl butan-1-amine, diisoamylamin, di-iso-amylamine, acmc-1astz
PubChem CID 10988
IUPAC Name 3-methyl-N-(3-methylbutyl)butan-1-amine
SMILES CC(C)CCNCCC(C)C

Specifications

Melting Point -44°C
Boiling Point 187°C to 188°C
Flash Point 58°C (136°F)
Quantity 5 g
UN Number UN2733
Beilstein 1719663
Merck Index 14,3193
Formula Weight 157.3
Percent Purity 97%
Chemical Name or Material Diisopentylamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.