missing translation for 'onlineSavingsMsg'
Learn More

Diisopentyl ether, mixture of isomers, 96%, Thermo Scientific Chemicals

Catalog Number 11444310
Click to view available options
Quantity:
25 mL
100 mL
500 mL
This item is not returnable. View return policy
544-01-4
C10H22O
158.285
MFCD00008947
AQZGPSLYZOOYQP-UHFFFAOYSA-N
diisopentyl ether, isoamyl ether, isopentyl ether, diisoamyl ether, isoamyl oxide, butane, 1,1'-oxybis 3-methyl, di-3-methylbutyl ether, di-iso-amyl ether, 1,1'-oxybis 3-methylbutane, 1,1'-oxylbis 3-methylbutane
10989
3-methyl-1-(3-methylbutoxy)butane
CC(C)CCOCCC(C)C
This item is not returnable. View return policy
544-01-4
C10H22O
158.285
MFCD00008947
AQZGPSLYZOOYQP-UHFFFAOYSA-N
diisopentyl ether, isoamyl ether, isopentyl ether, diisoamyl ether, isoamyl oxide, butane, 1,1'-oxybis 3-methyl, di-3-methylbutyl ether, di-iso-amyl ether, 1,1'-oxybis 3-methylbutane, 1,1'-oxylbis 3-methylbutane
10989
3-methyl-1-(3-methylbutoxy)butane
CC(C)CCOCCC(C)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.78
Boiling Point 172°C to 174°C
Flash Point 46°C (115°F)
Refractive Index 1.407
Quantity 25 mL
UN Number UN3271
Beilstein 1698014
Merck Index 14,5118
Formula Weight 158.29
Percent Purity 96%
Chemical Name or Material Diisopentyl ether, Mixture of isomers
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'