missing translation for 'onlineSavingsMsg'
Learn More

Diisopentyl ether, mixture of isomers, 96%

Product Code. 11454310
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454310 100 mL 100mL
11444310 25 mL 25mL
11464310 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11454310 Supplier Thermo Scientific Alfa Aesar Supplier No. A11231.AE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 544-01-4 | C10H22O | 158.285 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544-01-4
Molecular Formula C10H22O
Molecular Weight (g/mol) 158.285
MDL Number MFCD00008947
InChI Key AQZGPSLYZOOYQP-UHFFFAOYSA-N
Synonym diisopentyl ether, isoamyl ether, isopentyl ether, diisoamyl ether, isoamyl oxide, butane, 1,1'-oxybis 3-methyl, di-3-methylbutyl ether, di-iso-amyl ether, 1,1'-oxybis 3-methylbutane, 1,1'-oxylbis 3-methylbutane
PubChem CID 10989
IUPAC Name 3-methyl-1-(3-methylbutoxy)butane
SMILES CC(C)CCOCCC(C)C

Specifications

Density 0.78
Boiling Point 172°C to 174°C
Flash Point 46°C (115°F)
Refractive Index 1.407
Quantity 100 mL
UN Number UN3271
Beilstein 1698014
Merck Index 14,5118
Formula Weight 158.29
Percent Purity 96%
Chemical Name or Material Diisopentyl ether, Mixture of isomers
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.