missing translation for 'onlineSavingsMsg'
Learn More

Diisononyl Phthalate (mixture of branched chain isomers), TRC

Product Code. 30381341
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381341 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30381341 Supplier Toronto Research Chemicals Supplier No. TRCD455395100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diisononyl Phthalate (mixture of branched chain isomers)
CAS 68515-48-0
Molecular Formula C26H42O4
Quantity 100 g
Synonym 1,2-Benzenedicarboxylic acid, di-C8-10-branched alkyl esters, C9-rich, Phthalic acid, bis-iso-nonyl ester, Phthalic acid bis-isononyl ester, Di(C8-10-branched,C9-rich alkyl) phthalate
InChI Formula InChI=1S/C26H42O4/c1-3-5-7-9-11-13-17-21-29-25(27)23-19-15-16-20-24(23)26(28)30-22-18-14-12-10-8-6-4-2/h15-16,19-20H,3-14,17-18,21-22H2,1-2H3
SMILES O=C(OCCCCCCCCC)C1=C(C(OCCCCCCCCC)=O)C=CC=C1
IUPAC Name dinonyl phthalate
Molecular Weight (g/mol) 418.61
Formula Weight 222.0892
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.