missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diisodecyl Phthalate (Mixture of Branched chain Isomers) (>80%), TRC
Specifications
Specifications
| Chemical Name or Material | Phthalic acid, bis-isodecyl ester |
| CAS | 26761-40-0 |
| Molecular Formula | C28 H46 O4 |
| Quantity | 25 mL |
| Synonym | 1,2-Benzenedicarboxylic acid, diisodecyl ester (9CI), Phthalic acid, diisodecyl ester (6CI,8CI), Isodecyl alcohol, phthalate (2:1) (8CI), Bis(isodecyl) phthalate, DIDP, Diisodecyl phthalate, Emkarate 1020, Hexaplas DIDP, Isodecyl phthalate, Jayflex DIDP, PX 120, Palatinol DIDP, Palatinol Z, Plasthall DIDP, Reomol DiDP, Sansocizer DIDP, Sicol 184, Vestinol DZ, (mixture of isomers) |
| InChI Formula | InChI=1S/C28H46O4/c1-5-16-24(4)18-11-9-15-22-32-28(30)26-20-13-12-19-25(26)27(29)31-21-14-8-6-7-10-17-23(2)3/h12-13,19-20,23-24H,5-11,14-18,21-22H2,1-4H3 |
| SMILES | CCCC(C)CCCCCOC(=O)c1ccccc1C(=O)OCCCCCCCC(C)C |
| IUPAC Name | 2-O-(6-methylnonyl) 1-O-(8-methylnonyl) benzene-1,2-dicarboxylate |
| Molecular Weight (g/mol) | 446.66 |
| Formula Weight | 446.3396 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?