missing translation for 'onlineSavingsMsg'
Learn More

Diisobutylaluminum hydride, 1M solution in toluene

Product Code. 15460837
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15460837 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 15460837 Supplier Thermo Scientific Alfa Aesar Supplier No. L14718.AE

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 1191-15-7 | C8H19Al

Diisobutylaluminum hydride reduces chiral ?-silyloxy esters to the corresponding enantiomerically-pure hydroxy aldehydes. I t is used in Pd-catalyzed reductive debromination of secondary alkyl bromides, O-Debenzylation and ring opening of perbenzylated furanosides and convenient in situ generation of HZrCp2Cl from ZrCp2Cl2 and DIBAL-H.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diisobutylaluminum hydride reduces chiral ɑ-silyloxy esters to the corresponding enantiomerically-pure hydroxy aldehydes. I t is used in Pd-catalyzed reductive debromination of secondary alkyl bromides, O-Debenzylation and ring open
TRUSTED_SUSTAINABILITY

Specifications

CAS 1191-15-7
Density 0.805
Boiling Point ∼110°C
Flash Point 4°C (40°F)
Molecular Formula C8H19Al
Linear Formula [(CH3)2CHCH2]2AlH
MDL Number MFCD00008928
Quantity 100 mL
UN Number UN3399
Beilstein 4123663
Sensitivity Moisture sensitive
Synonym DIBAH; DIBAL-H
Solubility Information Reacts with water.
Formula Weight 142.22
Chemical Name or Material Diisobutylaluminum hydride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.