missing translation for 'onlineSavingsMsg'
Learn More

Diisobutylaluminum hydride, 1M solution in hexane, Thermo Scientific™

Product Code. 15416495
Click to view available options
Quantity:
250 mL
1 L
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Reducing agent

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1191-15-7
MDL Number MFCD00008928

Specifications

Color Colorless
Density 0.701 g/mL
Flash Point −23°C (−10°F)
Linear Formula [(CH3)2CHCH2]2AlH
Quantity 250 mL
UN Number UN3399
Sensitivity Air and moisture sensitive
Solubility Information Miscible with hydrocarbon solvents.
Formula Weight 142.22
Physical Form Liquid
Chemical Name or Material Diisobutylaluminum hydride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'