Learn More
Diisobutylaluminium hydride, 1M solution in hexane, AcroSeal™, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 183798000
Code : 29
Additional Details : CAS Number : 1191-15-7 Weight : 0.56080kg Transport : UN number : 3394 Chem class : 4.2 Pack group : I
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid, Reduction reagentSpecifications
Diisobutylaluminium hydride | |
1191-15-7,110-54-3 | |
0.7010g/mL | |
AcroSeal™ Glass Bottle | |
[(CH3)2CHCH2]2AlH | |
800 mL | |
01,260; 02,140; 03,101; 04,158; 05,224; 06,198; 07,111; 08,173; 09,171; 10,149; 11,185; 12,191; 13,115; 14,138; 15,137; 16,27; 17,123 | |
15, 3212 | |
Solubility in water: reacts | |
142.22 |
1M Solution in Hexane | |
-70.0°C | |
−23°C | |
C8H19Al | |
MFCD00008928 | |
04, IV, 4400 | |
0.701 | |
DIBAL-H | |
142.22 | |
Solution |
Safety and Handling
- Diisobutylaluminium hydride, 1M solution in hexane
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Substance which\, in contact with water\, emits flammable gases Category 1
- Pyrophoric liquid Category 1
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 1 A
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H250-Catches fire spontaneously if exposed to air.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H304-May be fatal if swallowed and enters airways.
- H411-Toxic to aquatic life with long lasting effects.
- H314-Causes severe skin burns and eye damage.
- H336-May cause drowsiness or dizziness.
- H373-May cause damage to organs.
- H361f-Suspected of damaging fertility.
- EUH014-Reacts violently with water.
Precautionary Statement
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P231+P232-Handle under inert gas. Protect from moisture.
- P302+P334-IF ON SKIN: Immerse in cool water/wrap in wet bandages.
- P335+P334-Brush off loose particles from skin Immerse in cool water/wrap in wet bandages.
- P310-Immediately call a POISON CENTER/doctor/.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P273-Avoid release to the environment.
- P402+P404-Store in a dry place. Store in a closed container.
- P222-Do not allow contact with air.
Supplemental information
- MIXTURE LIST-Contains : Hexane, Diisobutylaluminum hydride
RUO – Research Use Only