missing translation for 'onlineSavingsMsg'
Learn More

Diisobutyl Phthalate, TRC

Product Code. 30386444
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30386444 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30386444 Supplier Toronto Research Chemicals Supplier No. TRCD45521010G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

CAS 84-69-5
Molecular Formula C16 H22 O4
Quantity 10 g
Synonym 1,2-Benzenedicarboxylic acid, 1,2-bis(2-methylpropyl) ester, 1,2-Benzenedicarboxylic acid, bis(2-methylpropyl) ester (9CI), Phthalic acid, diisobutyl ester (6CI,7CI,8CI), 1,2-Benzenedicarboxylic acid diisobutyl ester, 2-Methylpropyl phthalate, Bis(2-methylpropyl) o-phthalate, Bis(2-methylpropyl) phthalate, Di(2-methylpropyl) phthalate, Di(isobutyl) 1,2-benzenedicarboxylate, Di-iso-Butyl phthalate, Diisobutyl phthalate, Hexaplas M/1B, Isobutyl phthalate, NSC 15316, Palatinol IC, Reomol DiBP
InChI Formula InChI=1S/C16H22O4/c1-11(2)9-19-15(17)13-7-5-6-8-14(13)16(18)20-10-12(3)4/h5-8,11-12H,9-10H2,1-4H3
SMILES CC(C)COC(=O)c1ccccc1C(=O)OCC(C)C
IUPAC Name bis(2-methylpropyl) benzene-1,2-dicarboxylate
Molecular Weight (g/mol) 278.34
Formula Weight 278.1518
Percent Purity >95%
Chemical Name or Material Phthalic acid, bis-iso-butyl ester
Grade HPLC
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.