missing translation for 'onlineSavingsMsg'
Learn More

Diisobutyl Phthalate-d4, TRC

Product Code. 30387069
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387069 100 mg 100mg
30381905 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30387069 Supplier Toronto Research Chemicals Supplier No. TRCD455212100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phthalic acid, bis-iso-butyl ester D4
CAS 358730-88-8
Purity Grade Notes HPLC
Molecular Formula C16 2H4 H18 O4
Quantity 100 mg
Synonym 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, bis(2-methylpropyl) ester (9CI), Phthalic acid, bis-iso-butyl ester D4, Diisobutyl Phthalate-d4, Phthalic Acid Diisobutyl Ester (3,4,5,6)-D4, Phthalic Acid, Diisobutyl Ester D4 (phenyl D4)
InChI Formula InChI=1S/C16H22O4/c1-11(2)9-19-15(17)13-7-5-6-8-14(13)16(18)20-10-12(3)4/h5-8,11-12H,9-10H2,1-4H3/i5D,6D,7D,8D
SMILES [2H]c1c([2H])c([2H])c(C(=O)OCC(C)C)c(C(=O)OCC(C)C)c1[2H]
IUPAC Name bis(2-methylpropyl) 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate
Molecular Weight (g/mol) 282.37
Formula Weight 282.1769
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.