missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diisobutyl Adipate, TRC
Specifications
Specifications
| Chemical Name or Material | Bis(2-methylpropyl) adipate |
| CAS | 141-04-8 |
| Molecular Formula | C14 H26 O4 |
| Quantity | 5 mL |
| Synonym | Adipic acid, diisobutyl ester (6CI,7CI,8CI), Hexanedioic acid, bis(2-methylpropyl) ester (9CI), Adipic acid bis(2-methylpropyl) ester, Adipic acid di-iso-butyl ester, DI 4A, DIBA, Diisobutyl adipate, Ftaflex DIBA, Isobutyl adipate, KAK-DIBA, NSC 6343, Plasthall DIBA, Rhodisolv DIB, Vinycizer 40 |
| InChI Formula | InChI=1S/C14H26O4/c1-11(2)9-17-13(15)7-5-6-8-14(16)18-10-12(3)4/h11-12H,5-10H2,1-4H3 |
| SMILES | CC(C)COC(=O)CCCCC(=O)OCC(C)C |
| IUPAC Name | bis(2-methylpropyl) hexanedioate |
| Molecular Weight (g/mol) | 258.35 |
| Formula Weight | 258.1831 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?