missing translation for 'onlineSavingsMsg'
Learn More

Diiododifluoromethane, TRC

Product Code. 30384805
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30384805 500 mg
30369599 50 mg
30380831 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30384805 Supplier Toronto Research Chemicals Supplier No. TRCD455383500MG
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1184-76-5
Molecular Formula CF2I2
Molecular Weight (g/mol) 303.82
Synonym Difluorodiiodomethane
IUPAC Name difluoro(diiodo)methane
SMILES FC(F)(I)I

Specifications

Quantity 500 mg
InChI Formula InChI=1S/CF2I2/c2-1(3,4)5
Formula Weight 303.8057
Chemical Name or Material Diiododifluoromethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.