missing translation for 'onlineSavingsMsg'
Learn More

Diiodobis(triphenylphosphine)nickel(II)

Product Code. 11373507
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11373507 2 g 2g
11383507 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11373507 Supplier Thermo Scientific Alfa Aesar Supplier No. 018869.04
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 82308-17-6 | C36H30I2NiP2 | 837.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82308-17-6
Molecular Formula C36H30I2NiP2
Molecular Weight (g/mol) 837.09
MDL Number MFCD00015866
InChI Key CRLSSYRKSIEFOQ-UHFFFAOYSA-L
Synonym diiododbis triphenylphosphinenickel ii, diiodobis triphenylphosphine nickel ii, nii2 pph3 2, diiodonickel; triphenylphosphane
PubChem CID 6102125
IUPAC Name diiodonickel;triphenylphosphane
SMILES I[Ni++]I.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Brown
Linear Formula [(C6H5)3P]2NiI2
Quantity 2 g
Sensitivity Air and light sensitive
Formula Weight 837.08
Physical Form Crystalline Powder
Chemical Name or Material Diiodobis(triphenylphosphine)nickel(II)
compliance-icons compliance-icons
Product Identifier
  • Diiodobis(triphenylphosphine)nickel(II)
Signal Word
  • Danger
Hazard Category
  • Skin sensitiser Category 1
  • Carcinogenicity Category 1B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H350-May cause cancer.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P363-Wash contaminated clothing before reuse.
  • P405-Store locked up.
  • P501-Dispose of contents/container to accordance with local/regional/national/international regulations..

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.