missing translation for 'onlineSavingsMsg'
Learn More

Dihydroxybis(ammonium lactato)titanium(IV), 50% w/w aq. soln.

Product Code. 11345407
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11345407 1 kg 1kg
11396129 250 g 250g
11335407 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11345407 Supplier Thermo Scientific Alfa Aesar Supplier No. 042676.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Water soluble titanium complex | CAS: 65104-06-5

Dihydroxybis(ammonium lactato)titanium(IV) is used in the production of water soluble titanium complex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65104-06-5
MDL Number MFCD00072736
Synonym Titanium(IV) bis(ammonium lactato)dihydroxide

Specifications

Color Yellow
Density 1.21 g/mL
Flash Point 47°C (118°F)
Refractive Index 1.438
Quantity 1 kg
UN Number UN1993
Solubility Information Miscible with water.
Formula Weight 294.1
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Physical Form Liquid
Chemical Name or Material Dihydroxybis (ammonium lactato)titanium (IV)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.