missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dihydrotestosterone Competitive ELISA Kit

Product Code. 30245369
Click to view available options
:
96 Tests
This item is not returnable. View return policy

Product Code. 30245369

Brand: Invitrogen™ EEL221

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Dihydrotestosterone (DHT) is a biologically active metabolite of the hormone testosterone, formed primarily in the prostate gland, testes, hair follicles, and adrenal glands by the enzyme 5-alpha-reductase by means of reducing the alpha 4, 5 double-bond. Dihydrotestosterone belongs to the class of compounds called androgens, also commonly called androgenic hormones or testoids. DHT is thought to be approximately 30 times more potent than testosterone because of increased affinity to the androgen receptor. [National Center for Biotechnology Information. PubChem Compound Database; CID=10635].
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 39.06 to 2500 pg/mL
Assay Sensitivity 23.44 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum
For Use With (Equipment) Colorimetric Microplate Reader
Interassay CV <10
Intraassay CV <10
Kit Contents Micro ELISA Plate (Dismountable), Reference Standard, Concentrated Biotinylated Detection Ab(100x), Concentrated HRP Conjugate (100x), Reference Standard & Sample Diluent, Biotinylated Detection Ab Diluent, HRP Conjugate Diluent, Concentrated Wash Buffer(25x), Substrate Reagent, Stop Solution, Plate Sealer, Product Description, Certificate of Analysis
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma: 100 μL, Serum: 100 μL
Storage Requirements Refer to product documentation for component specific storage temperature.
Target Dihydrotestosterone
Test Time 1 hr. 20 min.
Total Assay Time 3 hr. 30 min.
Vis mere Vis mindre
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.