missing translation for 'onlineSavingsMsg'
Learn More

Dihydrotanshinone I, MedChemExpress

Product Code. 16484817
Encompass_Preferred
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
This item is not returnable. View return policy

Product Code. 16484817

Brand: Medchem Express HYN0360

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dihydrotanshinone I is a natural compound extracted from Salvia miltiorrhiza Bunge which has been widely used for treating cardiovascular diseases. Dihydrotanshinone I exhibits entry-blocking effect for MERS-CoV.

Dihydrotanshinone I is a natural compound extracted from Salvia miltiorrhiza Bunge which has been widely used for treating cardiovascular diseases. Dihydrotanshinone I exhibits entry-blocking effect for MERS-CoV.

Specifications

Chemical Name or Material Dihydrotanshinone I
CAS 87205-99-0
Color Red
Purity Grade Notes Research
Molecular Formula C18H14O3
Quantity 50 mg
Solubility Information DMSO : 2 mg/mL (7.19 mM; Need ultrasonic)
SMILES O=C(C1=C2C=CC3=C1C=CC=C3C)C(C4=C2OC[C@@H]4C)=O
Molecular Weight (g/mol) 278.3
Formula Weight 278.3
Percent Purity 98.74%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.