missing translation for 'onlineSavingsMsg'
Learn More

Dihydrostreptomycin sulfate, Thermo Scientific™

Product Code. 15845128 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Plastic bottle
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15845128 100 g Plastic bottle 100g
15875278 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15845128 Supplier Thermo Scientific Supplier No. J14765.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5490-27-7
Molecular Formula C42H88N14O36S3
Molecular Weight (g/mol) 1461.408
MDL Number MFCD00070252
InChI Key CZWJCQXZZJHHRH-UHFFFAOYSA-N
Synonym dihydrostreptomycin sulfate, bis dihydrostreptomycin ; tris sulfuric acid
PubChem CID 44134677
IUPAC Name 2-[3-(diaminomethylideneamino)-4-[3-[4,5-dihydroxy-6-(hydroxymethyl)-3-(methylamino)oxan-2-yl]oxy-4-hydroxy-4-(hydroxymethyl)-5-methyloxolan-2-yl]oxy-2,5,6-trihydroxycyclohexyl]guanidine;sulfuric acid
SMILES CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(CO)O.CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(CO)O.OS(=O)(=O)O.OS(=O)(=O)O.OS(=O)(=O)O

Specifications

Packaging Plastic bottle
Quantity 100 g
Beilstein 3894221
Formula Weight 1461.42
Physical Form Powder
Chemical Name or Material Dihydrostreptomycin Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.