missing translation for 'onlineSavingsMsg'
Learn More

(-)-Dihydroquinine, TRC

Product Code. 30381635
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30381635 1 g
30386591 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30381635 Supplier Toronto Research Chemicals Supplier No. TRCD4499151G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 522-66-7
Molecular Formula C20 H26 N2 O2
Molecular Weight (g/mol) 326.43
Synonym Cinchonan-9-ol, 10,11-dihydro-6'-methoxy-, (8α,9R)-, Hydroquinine (6CI,7CI,8CI), (-)-10,11-Dihydroquinine, (-)-Dihydroquinine, (-)-Hydroquinine, (8α,9R)-10,11-Dihydro-6'-methoxycinchonan-9-ol, 10,11-Dihydroquinine, Dihydroquinine, NSC 41799
IUPAC Name (R)-[(2S,4S,5R)-5-ethyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol
SMILES CC[C@H]1CN2CC[C@H]1C[C@H]2[C@H](O)c3ccnc4ccc(OC)cc34

Specifications

Quantity 1 g
InChI Formula InChI=1S/C20H26N2O2/c1-3-13-12-22-9-7-14(13)10-19(22)20(23)16-6-8-21-18-5-4-15(24-2)11-17(16)18/h4-6,8,11,13-14,19-20,23H,3,7,9-10,12H2,1-2H3/t13-,14-,19-,20+/m0/s1
Formula Weight 326.1994
Chemical Name or Material (-)-Dihydroquinine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.