missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Dihydrolipoic Acid
Description
- Dihydrolipoic Acid (DHLA) is the reduced form of alpha-lipoic acid, and is a carboxylic acid containing two thiol groups (Moini et al.) It acts as a general antioxidant that is highly reactive against a variety of reactive oxygen species (ROS), including hydroxyl radicals, peroxynitrite, hydrogen peroxide, and hypochlorite, at concentrations ranging from 0.01 - 0.5 mM (Moini et al.) Dihydrolipoic acid has also been shown to recycle ubiquinone to the antioxidant active divalently reduced form (Nohl & Gille)
- Conversely, at concentrations higher than 50 - 100 μM, Dihydrolipoic acid directly increases the ROS content, along with a significant increase in cytoplasmic free calcium and nitric oxide (NO) levels, loss of mitochondrial membrane potential, activation of caspases-9 and -3, and cell death (Chan et al.; Houng et al.)
Specifications
Specifications
| CAS | 462-20-4 |
| Quantity | 50 mg |
| Molecular Formula | C8H16O2S2 |
| Purity | ≥95% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?