missing translation for 'onlineSavingsMsg'
Learn More

Dihydrogen hexahydroxyplatinate(IV), 99.9% (metals basis), Pt 61.0% min

Product Code. 11381698
Change view
Click to view available options
Quantity:
0.5 g
1 g
5 g
Unit Size:
0.5g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11381698 5 g 5g
11301708 0.5 g 0.5g
11391698 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11381698 Supplier Thermo Scientific Alfa Aesar Supplier No. 011052.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 51850-20-5 | H8O6Pt | 299.14 g/mol

Dihydrogen hexahydroxyplatinate(IV) is used as a precursor to produce platinum dioxide (II), a catalyst of hydrogenation and hydrogenolysis of carbonyl group or C-Cl linkages in acyl chlorides. It also acts as a precursor to produce other platinum compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51850-20-5
Molecular Formula H8O6Pt
Molecular Weight (g/mol) 299.14
MDL Number MFCD00058744
InChI Key ILXDKMTYQJUXAB-UHFFFAOYSA-J
Synonym Hexahydroxyplatinic acid; Hydrogen hexahydroxyplatinate(IV)
IUPAC Name platinum(4+) dihydrogen hexahydroxide
SMILES [H+].[H+].[OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[Pt+4]

Specifications

Melting Point (decomposition)
Color Yellow
Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Insoluble in water.
Formula Weight 299.17
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Pt ≤61.0%
Physical Form Crystalline
Assay (metals basis)
Chemical Name or Material Dihydrogen hexahydroxyplatinate(IV)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.