missing translation for 'onlineSavingsMsg'
Learn More

Dihydrogen hexafluorozirconate, 45% w/w aq. soln., Thermo Scientific Chemicals

Product Code. 11328268
Change view
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11328268 500 g 500g
11338268 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11328268 Supplier Thermo Scientific Alfa Aesar Supplier No. 039462.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Dihydrogen hexafluorozirconate is used in manufacturing of optical glass and fluozirconate. It acts as a precursor to zirconium dioxide ceramic thin films. Further, it is used for surface pretreatments on steel to prevent corrosion. In addition to this, it is used in the production of the atomic energy industry, advanced electrical materials and refractories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12021-95-3
Molecular Formula F6H2Zr
Molecular Weight (g/mol) 207.23
MDL Number MFCD00082965
InChI Key PZFQYBOZXGWNAM-UHFFFAOYSA-H
Synonym Hexafluorozirconic acid; Hydrogen hexafluorozirconate

Specifications

Color Colorless
Density 1.51g/mL
Quantity 500 g
UN Number UN2922
Formula Weight 207.22
Concentration or Composition (by Analyte or Components) 45% w/w aq. soln.
Physical Form Liquid
Chemical Name or Material Dihydrogen hexafluorozirconate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.