missing translation for 'onlineSavingsMsg'
Learn More

Dihydrogen hexachloroplatinate(IV) solution, Pt 20% (cont. Pt)

Product Code. 11420251
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420251 1 Ea. 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11420251 Supplier Thermo Scientific Alfa Aesar Supplier No. 040177.NC
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 16941-12-1 | H{2}Cl{6}Pt

Dihydrogen hexachloroplatinate(IV) is used as a catalyst in the determination of potassium and in the purification of platinum. It is also used in catalysis reactions, electroplating, photography, platinum mirrors, printing for etching of zinc, producing fine color in glass and porcelain, indelible ink and in microscopy. In the presence of copper(II) chloride, it is used to catalyze the photooxygenation of alcohols to aldehydes or ketones. It plays an important role in intramolecular hydrosilylation of an alkyne and hydrosilylation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifikationer

CAS 16941-12-1
Physical Form Liquid
Quantity 1 Ea.
Molecular Formula H{2}Cl{6}Pt
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.