Learn More
Dihydrogen hexachloroplatinate(IV) solution, Pt 20% (cont. Pt)
Description
Dihydrogen hexachloroplatinate(IV) is used as a catalyst in the determination of potassium and in the purification of platinum. It is also used in catalysis reactions, electroplating, photography, platinum mirrors, printing for etching of zinc, producing fine color in glass and porcelain, indelible ink and in microscopy. In the presence of copper(II) chloride, it is used to catalyze the photooxygenation of alcohols to aldehydes or ketones. It plays an important role in intramolecular hydrosilylation of an alkyne and hydrosilylation reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Red to Brown |
| Physical Form | Liquid |
| Quantity | 2 g |
| Linear Formula | H2PtCl6 |
| UN Number | UN3264 |
| Merck Index | 14,7526 |
| Solubility Information | Miscible with water,alcohol,acetone,ethyl acetate and ether. |
| Formula Weight | 409.82 (Anhydrous) |
| Concentration or Composition (by Analyte or Components) | Pt 20% (cont. Pt) |
| Chemical Name or Material | Dihydrogen hexachloroplatinate(IV) solution |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.