missing translation for 'onlineSavingsMsg'
Learn More

Dihydrogen hexachloroplatinate(IV) hydrate, Premion™, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11300288
Click to view available options
Quantity:
1 g
5 g

Dihydrogen hexachloroplatinate(IV) hydrate is used in the production of platinum metal and its compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26023-84-7
Molecular Formula Cl6H2Pt
Molecular Weight (g/mol) 409.80
MDL Number MFCD00149909
InChI Key ZKOQTCCVSRSECD-UHFFFAOYSA-J
Synonym Hexachloroplatinic acid hydrate
IUPAC Name dihydrogen hexachloroplatinumtetrakis(ylium)
SMILES [H+].[H+].Cl[Pt+4](Cl)(Cl)(Cl)(Cl)Cl

Specifications

Melting Point 60°C (decomposition)
Color Red to Brown
Assay Percent Range 99.999% (metals basis)
Odor Odorless
Quantity 1 g
Sensitivity Hygroscopic
Solubility Information Soluble in water,alcohol,acetone,ethyl acetate and ether.
Formula Weight 409.82 (Anhydrous)
Physical Form Crystals
Chemical Name or Material Dihydrogen hexachloroplatinate(IV) hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'