missing translation for 'onlineSavingsMsg'
Learn More

Dihydrogen hexachloroiridate(IV) hydrate, 99% (metals basis), Ir 38-42%

Product Code. 11321658
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11321658 5 g 5g
11311658 1 g 1g
11301658 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11321658 Supplier Thermo Scientific Alfa Aesar Supplier No. 011031.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 110802-84-1 | Cl6H2Ir | 406.93 g/mol

Dihydrogen hexachloroiridate(IV) hydrate is involved in the electrochemical synthesis of polyaniline on the surface of non-noble metal electrodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110802-84-1
Molecular Formula Cl6H2Ir
Molecular Weight (g/mol) 406.93
MDL Number MFCD00149906
InChI Key IBMVGQBDTOACBV-UHFFFAOYSA-J
Synonym Hydrogen hexachloroiridate(IV); Chloroiridic acid
PubChem CID 16211483
SMILES [H+].[H+].Cl[Ir+4](Cl)(Cl)(Cl)(Cl)Cl

Specifications

Color Black
Physical Form Lump
Quantity 5 g
UN Number UN3260
Solubility Information Soluble in alcohol and water
IUPAC Name dihydrogen hexachloroiridiumtetrakis(ylium)
Formula Weight 406.93 (Anhydrous)
Percent Purity 99%
Concentration or Composition (by Analyte or Components) Ir 38 to 42%
Assay (metals basis)
Chemical Name or Material Dihydrogen hexachloroiridate(IV) hydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dihydrogen hexachloroiridate(IV) hydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Dihydrogen hexachloroiridate(IV) hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.