missing translation for 'onlineSavingsMsg'
Learn More

Dihydrogen hexabromoplatinate(IV) nonahydrate, 99.9% (metals basis), Pt 23.7% min

Product Code. 11377467
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11377467 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11377467 Supplier Thermo Scientific Alfa Aesar Supplier No. 012617.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 20596-34-3 | Br6H2Pt | 676.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20596-34-3
Molecular Formula Br6H2Pt
Molecular Weight (g/mol) 676.52
MDL Number MFCD00049645
InChI Key VOEHEFXBCNCSCK-UHFFFAOYSA-J
Synonym Bromoplatinic acid; Platinic bromide
IUPAC Name platinum(4+) dihydrogen hexabromide
SMILES [H+].[H+].[Br-].[Br-].[Br-].[Br-].[Br-].[Br-].[Pt+4]

Specifications

Color Red to Brown
Linear Formula H2PtBr6·9H2O
Quantity 5 g
UN Number UN3260
Formula Weight 838.67 (676.56 Anhydrous)
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Pt 23.7% min.
Physical Form Crystalline
Assay (metals basis)
Chemical Name or Material Dihydrogen hexabromoplatinate(IV) nonahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.