missing translation for 'onlineSavingsMsg'
Learn More

Dihydrogen hexabromoosmate(IV) hydrate, 99.9% (metals basis)

Product Code. 11359108
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
0.25g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11359108 1 g 1g
11369108 0.25 g 0.25g
11349108 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11359108 Supplier Thermo Scientific Alfa Aesar Supplier No. 042527.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD01863474
Synonym Bromoosmic Acid; Hydrogen hexabromoosmate(IV)

Specifications

Melting Point (decomposition)
Color Gray to Black
Quantity 1 g
UN Number UN3260
Formula Weight 352.02 (Anhydrous)
Percent Purity 99.9%
Physical Form Crystalline
Assay (metals basis)
Chemical Name or Material Dihydrogen hexabromoosmate (IV) hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.