missing translation for 'onlineSavingsMsg'
Learn More

Dihydrogen dinitrosulfatoplatinate(II) solution, Pt 4-6% (cont. Pt)

Product Code. 11440300
Click to view available options
Quantity:
∼0.5 g
∼10 g
∼2 g
Unit Size:
0.5g
10g
2g
This item is not returnable. View return policy

Product Code. 11440300

Brand: Thermo Scientific Alfa Aesar 012627.NC

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12033-81-7 | H2N2O8PtS | 385.16 g/mol

Dihydrogen dinitrosulfatoplatinate(II) solution is used in the preparation of electrodeposition of platinum films and biocompatible platinum thin film electrodeposition on tin substrates, which acts as the interconnects material for implantable medical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12033-81-7
Molecular Formula H2N2O8PtS
Molecular Weight (g/mol) 385.16
MDL Number MFCD00064668
InChI Key SPPCQWREVYSZTJ-UHFFFAOYSA-N
Synonym dihydrogen dinitrosulfatoplatinate ii, dinitroplatinum; sulfuric acid, dihydrogen dinitrosulfatoplatinate ii solution, pt cont. pt
PubChem CID 102602532
IUPAC Name platinum;sulfuric acid;dinitrite
SMILES [H+].[H+].[O-]S([O-])(=O)=O.[O-][N+](=O)[Pt++][N+]([O-])=O

Specifications

Color Brown
Physical Form Liquid
Quantity ∼10 g
Solubility Information Miscible with water.
Formula Weight 384.17 (anhydrous)
Appearance Brown liquid
Chemical Name or Material Dihydrogen dinitrosulfatoplatinate(II) solution, Pt 4-6% (cont. Pt)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.