missing translation for 'onlineSavingsMsg'
Learn More

dihydrofuran-3(2h)-one, 95%, Maybridge

Brand:  Maybridge MO08444DE

Code : 50

Additional Details : CAS Number : 22929-52-8 Weight : 0.05500kg

Quantity 5g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10430954

  • £684.00 / 5g


Estimated Shipment date
from Supplier 29-06-2018
Add to basket

Specifications

Chemical Identifiers

Dihydrofuran-3(2H)-one
95.0
95%
86.09
22929-52-8
95%
C4H6O2
MFCD07778393

Specifications

Dihydrofuran-3(2H)-one
22929-52-8
Amber Glass Bottle
95%
86.09
5g
95.0
95%
C4H6O2
MFCD07778393
Videos
SDS