missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dihydrofolate Reductase/DHFR Antibody (SR1325), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21772-100ul
This item is not returnable.
View return policy
Description
Dihydrofolate Reductase/DHFR Monoclonal antibody specifically detects Dihydrofolate Reductase/DHFR in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Specifications
| Dihydrofolate Reductase/DHFR | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| SR1325 | |
| Western Blot 1:1000-1:5000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| DHFRP1, dihydrofolate reductase, DYR, EC 1.5.1.3 | |
| A synthesized peptide derived from human Dihydrofolate Reductase/DHFR (Uniprot #: P00374) | |
| 100 μg | |
| Cancer | |
| 1719 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction