missing translation for 'onlineSavingsMsg'
Learn More

Dihydrofolate Reductase/DHFR Antibody [PE], Novus Biologicals™

Product Code. 30733869 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30733869 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733869 Supplier Novus Biologicals Supplier No. AF7934P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Dihydrofolate Reductase/DHFR Polyclonal antibody specifically detects Dihydrofolate Reductase/DHFR in Human samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dihydrofolate Reductase/DHFR
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate PE
Formulation PBS
Gene Alias DHFRP1, dihydrofolate reductase, DYR, EC 1.5.1.3
Host Species Sheep
Immunogen E. coli-derived recombinant human Dihydrofolate Reductase/DHFR, Met1-Asp187, Accession # P00374
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1719
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.