missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dihydrofolate Reductase/DHFR Antibody (OTI6G7), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70569JF646
This item is not returnable.
View return policy
Description
Dihydrofolate Reductase/DHFR Monoclonal specifically detects Dihydrofolate Reductase/DHFR in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Dihydrofolate Reductase/DHFR | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| DHFR | |
| Full-length protein expressed in 293T cell transfected with human Dihydrofolate Reductase/DHFR expression vector. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI6G7 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| DHFRP1, dihydrofolate reductase, DYR, EC 1.5.1.3 | |
| Mouse | |
| Affinity Purified | |
| Cancer | |
| 1719.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction