missing translation for 'onlineSavingsMsg'
Learn More

Dihydrofolate Reductase/DHFR Antibody (OTI6G7), DyLight 594, Novus Biologicals™

Product Code. 18420113 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18420113 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18420113 Supplier Novus Biologicals Supplier No. NBP270569DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Dihydrofolate Reductase/DHFR Monoclonal antibody specifically detects Dihydrofolate Reductase/DHFR in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dihydrofolate Reductase/DHFR
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI6G7
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias DHFRP1, dihydrofolate reductase, DYR, EC 1.5.1.3
Host Species Mouse
Immunogen Full-length protein expressed in 293T cell transfected with human Dihydrofolate Reductase/DHFR expression vector.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1719
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.