missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dihydrofolate Reductase/DHFR Antibody (9B2), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-15195
This item is not returnable.
View return policy
Description
Dihydrofolate Reductase/DHFR Monoclonal antibody specifically detects Dihydrofolate Reductase/DHFR in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation
Specifications
| Dihydrofolate Reductase/DHFR | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150mM NaCl, and 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation | |
| 9B2 | |
| Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200, Immunoprecipitation | |
| DHFRP1, dihydrofolate reductase, DYR, EC 1.5.1.3 | |
| A synthesized peptide derived from human Dihydrofolate Reductase/DHFR. UniProt ID (P00374). | |
| 100 μg | |
| Cancer | |
| 1719 | |
| Store at -20 to -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction