missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dihydrofolate Reductase/DHFR Antibody (5B2), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Bio-Techne NBP2-59464H
This item is not returnable.
View return policy
Description
Dihydrofolate Reductase/DHFR Monoclonal antibody specifically detects Dihydrofolate Reductase/DHFR in Human samples. It is validated for Western Blot, ELISA
Specifications
| Dihydrofolate Reductase/DHFR | |
| Monoclonal | |
| HRP | |
| DHFRP1, dihydrofolate reductase, DYR, EC 1.5.1.3 | |
| Recombinant human Dihydrofolate Reductase/DHFR (1-187aa) purified from E. coli | |
| 0.1 mL | |
| Cancer | |
| 1719 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA | |
| 5B2 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction