missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Dihydroethidium

Product Code. 18075562
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18075562 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18075562 Supplier Tocris Bioscience™ Supplier No. 7642/25

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Fluorescent ROS indicator

Dihydroethidium is a fluorescent reactive oxygen species indicator. Cell permeable and suitable for use in live cells. Can be used with Mito-HE (Cat. No. 7641) to measure superoxide in living cells. Excitation/emission maxima λ 350/595 nm
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Ethyl-5,6-dihydro-6-phenyl-3,8-phenanthridinediamine
CAS 104821-25-2
Quantity 25 mg
Target Fluorescent Cell Indicators and Sensors
Molecular Formula C21H21N3
Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.